Product Details

SNP ID
rs138634383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49969750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCACCCTCAGGAACTCGAGGC[A/G]GGTAAGTTCGGCCGTGGAGGCTCTT
Phenotype
MIM: 601911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLX4 PubMed Links

Gene Details

Gene
DLX4
Gene Name
distal-less homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001934.3 206 Intron NP_001925.2
NM_138281.2 206 Silent Mutation GCA,GCG A94A NP_612138.1
XM_017024291.1 206 UTR 5 XP_016879780.1

View Full Product Details