Product Details

SNP ID
rs138749305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8141840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCCGAGGCAGTTGGCCCTTCC[A/G]GACCCAGGAGTGCACAGCACCCAGT
Phenotype
MIM: 602260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6883 PubMed Links

Gene Details

Gene
MIR6883
Gene Name
microRNA 6883
There are no transcripts associated with this gene.

Gene
PER1
Gene Name
period circadian clock 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002616.2 3623 Missense Mutation NP_002607.2
XM_005256689.1 3623 Missense Mutation XP_005256746.1
XM_005256690.1 3623 Missense Mutation XP_005256747.1

View Full Product Details