Product Details

SNP ID
rs138932612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81845190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCACCGCTCTCCAGGAATTTC[C/T]TAAAACCATCCAGCGTGCGTTCCCC
Phenotype
MIM: 176790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HB PubMed Links

Gene Details

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000918.3 1597 Missense Mutation AAG,AGG K467R NP_000909.2

View Full Product Details