Product Details

SNP ID
rs139483184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36013233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTTGATCCGTGTGTCCAGCTT[C/T]AGCATTCTCATGCAAACCTGAACTT
Phenotype
MIM: 602494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 449 Silent Mutation CTA,CTG L126L NP_005055.3
NM_145898.3 449 Silent Mutation CTA,CTG L109L NP_665905.2

View Full Product Details