Product Details

SNP ID
rs139491020
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18263238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTTGCCCTTGGCCCCCTCGTC[A/G]TCTGCCCCAGGTGAGTGGCACTCCT
Phenotype
MIM: 600362 MIM: 615498 MIM: 601243
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FLII PubMed Links

Gene Details

Gene
FLII
Gene Name
FLII, actin remodeling protein
There are no transcripts associated with this gene.

Gene
MIEF2
Gene Name
mitochondrial elongation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144900.1 511 Silent Mutation TCA,TCG S100S NP_001138372.1
NM_139162.3 511 Silent Mutation TCA,TCG S100S NP_631901.2
NM_148886.1 511 Silent Mutation TCA,TCG S111S NP_683684.2
XM_017024190.1 511 Silent Mutation TCA,TCG S107S XP_016879679.1
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
There are no transcripts associated with this gene.

View Full Product Details