Product Details

SNP ID
rs139649633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8369336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAAAGTAGCCACAGTTTCCCGG[G/T]GCAAGTTTGAGGAATATAGCCCAAA
Phenotype
MIM: 603704
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304947.1 2529 Missense Mutation CAC,CCC H344P NP_001291876.1
NM_213597.3 2529 Missense Mutation CAC,CCC H426P NP_998762.1
XM_005256443.2 2529 Missense Mutation CAC,CCC H392P XP_005256500.1
XM_011523643.1 2529 Missense Mutation CAC,CCC H392P XP_011521945.1
XM_011523644.1 2529 Missense Mutation CAC,CCC H392P XP_011521946.1
XM_017024163.1 2529 Missense Mutation CAC,CCC H392P XP_016879652.1
XM_017024164.1 2529 Missense Mutation CAC,CCC H389P XP_016879653.1
XM_017024165.1 2529 Missense Mutation CAC,CCC H344P XP_016879654.1
Gene
LOC100128288
Gene Name
uncharacterized LOC100128288
There are no transcripts associated with this gene.

Gene
RPL26
Gene Name
ribosomal protein L26
There are no transcripts associated with this gene.

View Full Product Details