Product Details

SNP ID
rs139838601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42612495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGGTTCCTACCTCTTAGAACG[A/G]CTGAATGACAGGTAAGTTTGTGTTT
Phenotype
MIM: 616498 MIM: 191135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM134C PubMed Links

Gene Details

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
There are no transcripts associated with this gene.

Gene
TUBG1
Gene Name
tubulin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001070.4 866 Silent Mutation CGA,CGG R156R NP_001061.2

View Full Product Details