Product Details
- SNP ID
-
rs139900511
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:10445043 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCAGCTGCTCAGCCTCATCCAGA[C/T]GGAGCTGCAGATCCTTCACGGTCTG
- Phenotype
-
MIM: 160742
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MYH4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2277649] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYH4
- Gene Name
- myosin heavy chain 4
- Gene
- MYHAS
- Gene Name
- myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.
View Full Product Details