Product Details

SNP ID
rs139919914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76266838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCGATAGGGATCTTAAAACA[C/T]TGTTCCCCAGGAGAGCTGTTGTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
QRICH2 PubMed Links

Gene Details

Gene
QRICH2
Gene Name
glutamine rich 2
There are no transcripts associated with this gene.

Gene
UBALD2
Gene Name
UBA like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182565.3 Intron NP_872371.1

View Full Product Details