Product Details

SNP ID
rs139962677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28554478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTCTGCCGACTTCTGGACGGC[A/G]GCTACAGCCTTGTACACCTAGGAAG
Phenotype
MIM: 610271 MIM: 604011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIGS PubMed Links

Gene Details

Gene
PIGS
Gene Name
phosphatidylinositol glycan anchor biosynthesis class S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033198.3 1783 Silent Mutation GCC,GCT A470A NP_149975.1
Gene
UNC119
Gene Name
unc-119 lipid binding chaperone
There are no transcripts associated with this gene.

View Full Product Details