Product Details

SNP ID
rs140618444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16623085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGTGTGAATTCTCTGATGTA[C/T]AGTTAAGTGTGATTTTATTCTGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF624 PubMed Links

Gene Details

Gene
ZNF624
Gene Name
zinc finger protein 624
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020787.3 1893 Missense Mutation ATA,GTA I601V NP_065838.2
XM_006721562.3 1893 Missense Mutation ATA,GTA I601V XP_006721625.1
XM_011523970.2 1893 Missense Mutation ATA,GTA I599V XP_011522272.1

View Full Product Details