Product Details

SNP ID
rs141034963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76716437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTCAATGAGACAGCACAGAAC[C/T]TTCTTACCCAACATCCCATTAGCTT
Phenotype
MIM: 604914 MIM: 615262
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD6 PubMed Links

Gene Details

Gene
JMJD6
Gene Name
arginine demethylase and lysine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081461.1 1818 UTR 3 NP_001074930.1
NM_015167.2 1818 Intron NP_055982.2
Gene
METTL23
Gene Name
methyltransferase like 23
There are no transcripts associated with this gene.

Gene
MXRA7
Gene Name
matrix remodeling associated 7
There are no transcripts associated with this gene.

View Full Product Details