Product Details
- SNP ID
-
rs141718604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:68271605 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATGTTTAACTGTAGTAGGCCCAC[A/G]AAGAGGAGGCTGTATCTCCAGCCAA
- Phenotype
-
MIM: 610008
MIM: 603880
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARSG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7222013] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARSG
- Gene Name
- arylsulfatase G
- Gene
- SLC16A6
- Gene Name
- solute carrier family 16 member 6
View Full Product Details