Product Details

SNP ID
rs141770749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50966122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTCTGTATTGTTATGGTAGAA[C/T]GGATTTTGTAATATAAAGTTAACAG
Phenotype
MIM: 605430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPAG9 PubMed Links

Gene Details

Gene
SPAG9
Gene Name
sperm associated antigen 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130527.2 4364 UTR 3 NP_001123999.1
NM_001130528.2 4364 UTR 3 NP_001124000.1
NM_001251971.1 4364 UTR 3 NP_001238900.1
NM_003971.5 4364 UTR 3 NP_003962.3
XM_005257768.2 4364 UTR 3 XP_005257825.1
XM_005257771.2 4364 UTR 3 XP_005257828.1
XM_005257774.3 4364 UTR 3 XP_005257831.1
XM_017025283.1 4364 UTR 3 XP_016880772.1
XM_017025284.1 4364 UTR 3 XP_016880773.1
XM_017025285.1 4364 UTR 3 XP_016880774.1

View Full Product Details