Product Details

SNP ID
rs141992223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63881141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTTCTGCTCCTTCAGGATATAG[G/T]CTTCTTCCTAGGAGAAGGACCGCCC
Phenotype
MIM: 118820 MIM: 139240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSH2 PubMed Links

Gene Details

Gene
CSH2
Gene Name
chorionic somatomammotropin hormone 2
There are no transcripts associated with this gene.

Gene
GH2
Gene Name
growth hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002059.4 320 Missense Mutation GAC,GCC D60A NP_002050.1
NM_022556.3 320 Intron NP_072050.1
NM_022557.3 320 Missense Mutation GAC,GCC D60A NP_072051.1
NM_022558.3 320 Missense Mutation GAC,GCC D60A NP_072052.1
Gene
LOC107987248
Gene Name
intercellular adhesion molecule 1-like
There are no transcripts associated with this gene.

View Full Product Details