Product Details

SNP ID
rs142447047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57840966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCGTCTTAAAATGACACCTTCT[A/G]CCAATAAAGCCTTCCCTGTTTCCAC
Phenotype
MIM: 611985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS23 PubMed Links

Gene Details

Gene
MRPS23
Gene Name
mitochondrial ribosomal protein S23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016070.3 442 Missense Mutation GCA,GTA A127V NP_057154.2

View Full Product Details