Product Details

SNP ID
rs142476209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82463735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGTCAGGAAGGAGAGGGTGGA[A/G]GGGATCAAAGAGTGACTCTACATCC
Phenotype
MIM: 605349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NARF PubMed Links

Gene Details

Gene
NARF
Gene Name
nuclear prelamin A recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038618.2 Intron NP_001033707.1
NM_001083608.1 Intron NP_001077077.1
NM_012336.3 Intron NP_036468.1
NM_031968.2 Intron NP_114174.1
XM_005256340.3 Intron XP_005256397.1
XM_006722278.3 Intron XP_006722341.1
XM_011523541.2 Intron XP_011521843.1
XM_011523543.2 Intron XP_011521845.1
XM_011523544.2 Intron XP_011521846.1
XM_017024449.1 Intron XP_016879938.1
XM_017024450.1 Intron XP_016879939.1

View Full Product Details