Product Details

SNP ID
rs142631346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36938212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAAAGACCATGGTTCACTGTGC[C/G]GGCTGCAAAAGGCCCATCCTGGACC
Phenotype
MIM: 601999
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LHX1 PubMed Links

Gene Details

Gene
LHX1
Gene Name
LIM homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005568.4 738 Silent Mutation GCC,GCG A5A NP_005559.2
Gene
LOC102723471
Gene Name
uncharacterized LOC102723471
There are no transcripts associated with this gene.

View Full Product Details