Product Details

SNP ID
rs142653327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27647416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCCGAAAAATGCCCTTCGTCC[A/G]TGGCCAGAGCTTCTCAGTAAGGCAC
Phenotype
MIM: 601879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGALS9 PubMed Links

Gene Details

Gene
LGALS9
Gene Name
galectin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002308.3 Intron NP_002299.2
NM_009587.2 Intron NP_033665.1
XM_006721892.2 Intron XP_006721955.1
XM_006721893.2 Intron XP_006721956.1
XM_006721894.3 Intron XP_006721957.1
XM_006721895.3 Intron XP_006721958.1
XM_011524796.1 Intron XP_011523098.1
XM_017024623.1 Intron XP_016880112.1

View Full Product Details