Product Details

SNP ID
rs142895125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41502476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTACTCTTGGTTCTGGCACTCC[A/G]TCTCACTGCGGAGCTCGCTCAGCTG
Phenotype
MIM: 148065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT13 PubMed Links

Gene Details

Gene
KRT13
Gene Name
keratin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002274.3 1205 Missense Mutation ACG,ATG T381M NP_002265.2
NM_153490.2 1205 Missense Mutation ACG,ATG T381M NP_705694.2

View Full Product Details