Product Details

SNP ID
rs143485066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82079588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACGCTCCTCTAGGCCCTTCAG[C/T]GGCAGCAGCGCCTCCAGCACCTGTG
Phenotype
MIM: 600212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FASN PubMed Links

Gene Details

Gene
FASN
Gene Name
fatty acid synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004104.4 7284 Silent Mutation CCA,CCG P2389P NP_004095.4
XM_011523538.2 7284 Silent Mutation CCA,CCG P2389P XP_011521840.1

View Full Product Details