Product Details

SNP ID
rs143865170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74941626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGAATGTGGTGTTCCTGGGT[A/G]GCGCCTTCATCTGCAGCATGATCTT
Phenotype
MIM: 605752 MIM: 607827 MIM: 607828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
There are no transcripts associated with this gene.

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
OTOP3
Gene Name
otopetrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272005.1 373 Missense Mutation AGC,GGC S85G NP_001258934.1
NM_178233.2 373 Missense Mutation AGC,GGC S103G NP_839947.1
XM_011524744.2 373 Missense Mutation AGC,GGC S74G XP_011523046.1

View Full Product Details