Product Details

SNP ID
rs143897454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40698626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTACAGATCCTGAGTTTCTACC[A/T]CCAAATTCTGCAAAACTAGAACCAC
Phenotype
MIM: 607742
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT24 PubMed Links

Gene Details

Gene
KRT24
Gene Name
keratin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019016.2 1119 Silent Mutation GGA,GGT G462G NP_061889.2
XM_006721739.3 1119 Silent Mutation GGA,GGT G364G XP_006721802.1
XM_011524460.2 1119 Silent Mutation GGA,GGT G359G XP_011522762.1
XM_017024299.1 1119 Intron XP_016879788.1
Gene
LOC105371775
Gene Name
uncharacterized LOC105371775
There are no transcripts associated with this gene.

View Full Product Details