Product Details

SNP ID
rs143988077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47847708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCCCCATCGGGCCCACATGGA[A/G]CCCCCAGTCGCTCCGCCTCCAGACA
Phenotype
MIM: 614966 MIM: 608613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC46 PubMed Links

Gene Details

Gene
LRRC46
Gene Name
leucine rich repeat containing 46
There are no transcripts associated with this gene.

Gene
SCRN2
Gene Name
secernin 2
There are no transcripts associated with this gene.

Gene
SP6
Gene Name
Sp6 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258248.1 963 Missense Mutation GCT,GTT A241V NP_001245177.1
NM_199262.2 963 Missense Mutation GCT,GTT A241V NP_954871.1
XM_006722115.3 963 Missense Mutation GCT,GTT A241V XP_006722178.1

View Full Product Details