Product Details

SNP ID
rs145240163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63697105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGACTTCTGCCTGTGTCCCAC[C/T]GGGGCGCCCTCAGGTCCTCCCAACT
Phenotype
MIM: 602539 MIM: 608626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIMD2 PubMed Links

Gene Details

Gene
LIMD2
Gene Name
LIM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030576.3 2009 UTR 3 NP_085053.1
XM_005257703.1 2009 Intron XP_005257760.1
XM_005257705.3 2009 Intron XP_005257762.1
XM_006722124.1 2009 Intron XP_006722187.1
Gene
LOC729683
Gene Name
uncharacterized LOC729683
There are no transcripts associated with this gene.

Gene
MAP3K3
Gene Name
mitogen-activated protein kinase kinase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002401.3 2009 Intron NP_002392.2
NM_203351.1 2009 Intron NP_976226.1
XM_005257376.3 2009 Intron XP_005257433.1
XM_005257377.3 2009 Intron XP_005257434.1
XM_005257378.2 2009 Intron XP_005257435.1
Gene
STRADA
Gene Name
STE20-related kinase adaptor alpha
There are no transcripts associated with this gene.

View Full Product Details