Product Details

SNP ID
rs145403333
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3397964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGCATGGAAGGTGGTCAGCAC[C/T]CAGGACAGCGCCACCAGGGCGAGAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR1E1 PubMed Links

Gene Details

Gene
OR1E1
Gene Name
olfactory receptor family 1 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003553.2 447 Nonsense Mutation TGA,TGG *149W NP_003544.2

View Full Product Details