Product Details

SNP ID
rs145490715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69079220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCACTTGACTTACCTTCTCCAGT[A/G]TGCACTGAGAAAGGCTGTATTCTTC
Phenotype
MIM: 612504 MIM: 612507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA6 PubMed Links

Gene Details

Gene
ABCA6
Gene Name
ATP binding cassette subfamily A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080284.2 5229 Missense Mutation ACA,ATA T1581I NP_525023.2
XM_011524574.2 5229 Missense Mutation ACA,ATA T1264I XP_011522876.1
XM_017024404.1 5229 Missense Mutation ACA,ATA T1581I XP_016879893.1
XM_017024405.1 5229 Missense Mutation ACA,ATA T1581I XP_016879894.1
XM_017024406.1 5229 Missense Mutation ACA,ATA T1386I XP_016879895.1
XM_017024407.1 5229 Missense Mutation ACA,ATA T1386I XP_016879896.1
XM_017024408.1 5229 Missense Mutation ACA,ATA T896I XP_016879897.1
Gene
ABCA9
Gene Name
ATP binding cassette subfamily A member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080283.3 5229 Intron NP_525022.2
XM_017024011.1 5229 Intron XP_016879500.1
XM_017024012.1 5229 Intron XP_016879501.1
XM_017024013.1 5229 Intron XP_016879502.1
XM_017024014.1 5229 Intron XP_016879503.1
XM_017024015.1 5229 Intron XP_016879504.1

View Full Product Details