Product Details

SNP ID
rs145685499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49206516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCCATGGGGCCAACCGCTC[A/G]GCCAGCCACACTGTCCTCAGAGTGT
Phenotype
MIM: 606363 MIM: 139391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABI3 PubMed Links

Gene Details

Gene
ABI3
Gene Name
ABI family member 3
There are no transcripts associated with this gene.

Gene
GNGT2
Gene Name
G protein subunit gamma transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198754.1 741 UTR 3 NP_001185683.1
NM_001198755.1 741 UTR 3 NP_001185684.1
NM_001198756.1 741 UTR 3 NP_001185685.1
NM_031498.2 741 UTR 3 NP_113686.1

View Full Product Details