Product Details

SNP ID
rs146160539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60156672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACGCTTCTTCTTCTCTGGCTA[C/T]GATAAGAAGCAAACGTGGACTGTCC
Phenotype
MIM: 114760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA4 PubMed Links

Gene Details

Gene
CA4
Gene Name
carbonic anhydrase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005257639.2 319 Silent Mutation TAC,TAT Y75Y XP_005257696.1
XM_011525183.2 319 UTR 5 XP_011523485.1
XM_017025012.1 319 UTR 5 XP_016880501.1

View Full Product Details