Product Details

SNP ID
rs146170394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41159979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGACAGCCTCAGCAGCAAGAA[A/G]CACTAGAGCAGGTTGGGCGGCAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-3 PubMed Links

Gene Details

Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
There are no transcripts associated with this gene.

Gene
KRTAP4-4
Gene Name
keratin associated protein 4-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032524.1 753 UTR 3 NP_115913.1

View Full Product Details