Product Details

SNP ID
rs146523753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79834058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGGCTTGAACTCGCTCAGCGA[C/T]TCTGCAGGTTCGTCCTGGGCCTCGG
Phenotype
MIM: 603079
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBX4 PubMed Links

Gene Details

Gene
CBX4
Gene Name
chromobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003655.2 3256 Silent Mutation GAA,GAG E528E NP_003646.2
XM_011525399.2 3256 Silent Mutation GAA,GAG E462E XP_011523701.1

View Full Product Details