Product Details
- SNP ID
-
rs146598881
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:61590618 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTGCCTTTGCTCTCGACACATTT[A/G]CTTGTGACATGACCAATTTCACGTC
- Phenotype
-
MIM: 609274
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NACA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61739273] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NACA2
- Gene Name
- nascent polypeptide associated complex alpha subunit 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199290.3 |
|
Intron |
|
|
NP_954984.1 |
View Full Product Details