Product Details
- SNP ID
-
rs146966502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75727260 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTCCGTGTGGATAAGGACTGC[A/G]CCTACTGCACAGACGAGGTGAGGAC
- Phenotype
-
MIM: 147557
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITGB4
PubMed Links
Gene Details
- Gene
- ITGB4
- Gene Name
- integrin subunit beta 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000213.4 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
NP_000204.3 |
NM_001005619.1 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
NP_001005619.1 |
NM_001005731.2 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
NP_001005731.1 |
NM_001321123.1 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
NP_001308052.1 |
XM_005257309.2 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
XP_005257366.1 |
XM_005257311.4 |
356 |
Missense Mutation |
ACC,GCC |
T49A |
XP_005257368.1 |
XM_006721866.3 |
356 |
Missense Mutation |
ACC,GCC |
T84A |
XP_006721929.1 |
XM_006721867.3 |
356 |
Missense Mutation |
ACC,GCC |
T84A |
XP_006721930.1 |
XM_006721868.3 |
356 |
Missense Mutation |
ACC,GCC |
T84A |
XP_006721931.1 |
XM_006721870.3 |
356 |
Missense Mutation |
ACC,GCC |
T84A |
XP_006721933.1 |
XM_011524751.2 |
356 |
Missense Mutation |
ACC,GCC |
T84A |
XP_011523053.1 |
XM_011524752.2 |
356 |
Intron |
|
|
XP_011523054.1 |
View Full Product Details