Product Details
- SNP ID
-
rs146991485
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48077159 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATAATATTGTATGCTGCTTATAG[C/T]AAGTGTTAGGCTTCTTCTCACACTC
- Phenotype
-
MIM: 604511
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBX1
PubMed Links
Gene Details
- Gene
- CBX1
- Gene Name
- chromobox 1
View Full Product Details