Product Details

SNP ID
rs147076627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48113352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCCTGTGTGCGGCGCCGCTAC[C/T]GTGAGTTCGTGTGGCTGAGAAAGCA
Phenotype
MIM: 614906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX11 PubMed Links

Gene Details

Gene
SNX11
Gene Name
sorting nexin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013323.2 448 Missense Mutation CGT,TGT R61C NP_037455.2
NM_152244.1 448 Missense Mutation CGT,TGT R61C NP_689450.1
XM_005257260.3 448 Missense Mutation CGT,TGT R61C XP_005257317.1
XM_005257261.3 448 Missense Mutation CGT,TGT R53C XP_005257318.1
XM_005257262.3 448 Missense Mutation CGT,TGT R53C XP_005257319.1
XM_011524697.2 448 Missense Mutation CGT,TGT R61C XP_011522999.1

View Full Product Details