Product Details

SNP ID
rs147197890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82080172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCATGTCCGTGAACTGCTGCA[C/T]GAAGAAGCATATGGCCTCCGTCTCA
Phenotype
MIM: 600212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FASN PubMed Links

Gene Details

Gene
FASN
Gene Name
fatty acid synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004104.4 7231 Missense Mutation ATG,GTG M2372V NP_004095.4
XM_011523538.2 7231 Missense Mutation ATG,GTG M2372V XP_011521840.1
Gene
SNORD134
Gene Name
small nucleolar RNA, C/D box 134
There are no transcripts associated with this gene.

View Full Product Details