Product Details

SNP ID
rs147394862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3291741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGCATGGGATTGATGACAGTG[C/T]TGAAAATTCCAACAGCTTTATCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR3A1 PubMed Links

Gene Details

Gene
OR3A1
Gene Name
olfactory receptor family 3 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002550.2 842 Missense Mutation NP_002541.2
XM_017024695.1 842 Intron XP_016880184.1
XM_017024696.1 842 Intron XP_016880185.1
Gene
OR3A2
Gene Name
olfactory receptor family 3 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002551.3 842 Intron NP_002542.3
XM_017024697.1 842 Intron XP_016880186.1
XM_017024698.1 842 Intron XP_016880187.1

View Full Product Details