Product Details

SNP ID
rs148232854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81665417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTCAGGTGGCAGCGATGCCTT[C/G]GGCGGCCTGGTGCCGTCCGCACCTG
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC137 PubMed Links

Gene Details

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
There are no transcripts associated with this gene.

Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039842.2 288 Silent Mutation CCC,CCG P76P NP_001034931.1
NM_001304994.1 288 Silent Mutation CCC,CCG P66P NP_001291923.1
NM_001304995.1 288 UTR 3 NP_001291924.1
NM_001304999.1 288 UTR 3 NP_001291928.1
Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details