Product Details

SNP ID
rs148239346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58521760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCGTTTCTTGTGGTCCACTTC[A/G]GGAGGTGTCAGTGTGTCTGCCTTAG
Phenotype
MIM: 603559 MIM: 603696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTMR4 PubMed Links

Gene Details

Gene
MTMR4
Gene Name
myotubularin related protein 4
There are no transcripts associated with this gene.

Gene
SEPT4
Gene Name
septin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198713.1 698 Silent Mutation NP_001185642.1
NM_001256782.1 698 Silent Mutation NP_001243711.1
NM_001256822.1 698 Silent Mutation NP_001243751.1
NM_004574.4 698 Silent Mutation NP_004565.1
NM_080415.3 698 UTR 3 NP_536340.1
NM_080416.3 698 Silent Mutation NP_536341.1
XM_006721949.2 698 Silent Mutation XP_006722012.1
XM_006721950.3 698 Silent Mutation XP_006722013.1
XM_006721951.2 698 Silent Mutation XP_006722014.1
XM_006721952.2 698 Silent Mutation XP_006722015.1
XM_006721954.2 698 Silent Mutation XP_006722017.1
XM_006721955.2 698 Silent Mutation XP_006722018.1
XM_011524911.1 698 Silent Mutation XP_011523213.1
XM_011524912.1 698 Silent Mutation XP_011523214.1
Gene
SEPT4-AS1
Gene Name
SEPT4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details