Product Details

SNP ID
rs148251573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42844782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCCTGGCCACACCCTGGCCA[A/G]AGCCAGCTGTTTGCAGACCTGAGCC
Phenotype
MIM: 602268 MIM: 603735 MIM: 605129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AOC2 PubMed Links

Gene Details

Gene
AOC2
Gene Name
amine oxidase, copper containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158.3 191 Silent Mutation CAA,CAG Q52Q NP_001149.2
NM_009590.2 191 Silent Mutation CAA,CAG Q52Q NP_033720.2
Gene
AOC3
Gene Name
amine oxidase, copper containing 3
There are no transcripts associated with this gene.

Gene
PSME3
Gene Name
proteasome activator subunit 3
There are no transcripts associated with this gene.

View Full Product Details