Product Details

SNP ID
rs148549870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8730174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTTCTTTTTCACCTCTGCCC[A/G]GATGTCCGACCGGTCTTGGATAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC42 PubMed Links

Gene Details

Gene
CCDC42
Gene Name
coiled-coil domain containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158261.1 912 Missense Mutation CGG,TGG R229W NP_001151733.1
NM_144681.2 912 Missense Mutation CGG,TGG R303W NP_653282.2

View Full Product Details