Product Details

SNP ID
rs148570791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82460024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAAAACAAAAACTGATGACCA[A/G]GAGAATGTGTCAGCCGATGCACCGA
Phenotype
MIM: 605349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf62 PubMed Links

Gene Details

Gene
C17orf62
Gene Name
chromosome 17 open reading frame 62
There are no transcripts associated with this gene.

Gene
NARF
Gene Name
nuclear prelamin A recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038618.2 106 UTR 5 NP_001033707.1
NM_001083608.1 106 Silent Mutation CAA,CAG Q20Q NP_001077077.1
NM_012336.3 106 Silent Mutation CAA,CAG Q20Q NP_036468.1
NM_031968.2 106 Silent Mutation CAA,CAG Q20Q NP_114174.1
XM_005256340.3 106 UTR 5 XP_005256397.1
XM_006722278.3 106 Silent Mutation CAA,CAG Q20Q XP_006722341.1
XM_011523541.2 106 UTR 5 XP_011521843.1
XM_011523543.2 106 UTR 5 XP_011521845.1
XM_011523544.2 106 Silent Mutation CAA,CAG Q20Q XP_011521846.1
XM_017024449.1 106 UTR 5 XP_016879938.1
XM_017024450.1 106 UTR 5 XP_016879939.1

View Full Product Details