Product Details

SNP ID
rs148666799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47848091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCACGAGCCATCCTCCGCGCC[C/T]GGGTGAGTCGGCCTGAACCACGATT
Phenotype
MIM: 614966 MIM: 608613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCRN2 PubMed Links

Gene Details

Gene
SCRN2
Gene Name
secernin 2
There are no transcripts associated with this gene.

Gene
SP6
Gene Name
Sp6 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258248.1 580 Silent Mutation CCA,CCG P113P NP_001245177.1
NM_199262.2 580 Silent Mutation CCA,CCG P113P NP_954871.1
XM_006722115.3 580 Silent Mutation CCA,CCG P113P XP_006722178.1

View Full Product Details