Product Details

SNP ID
rs148926603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41938680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGCCGATCTACGCCAGGGAGC[A/G]GGACCGGAAGCTGATGCAAGAGAAA
Phenotype
MIM: 108728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
There are no transcripts associated with this gene.

Gene
TTC25
Gene Name
tetratricopeptide repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031421.3 850 Missense Mutation CAG,CGG Q250R NP_113609.1
XM_011525323.2 850 Missense Mutation CAG,CGG Q87R XP_011523625.1
XM_011525324.2 850 Missense Mutation CAG,CGG Q250R XP_011523626.1

View Full Product Details