Product Details

SNP ID
rs149016816
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81821761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCCATGGGGGCCAGGTAAGGC[C/T]GTGTGTTTACAGGGAGTTGGCTCGG
Phenotype
MIM: 138033 MIM: 616514
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
There are no transcripts associated with this gene.

Gene
MCRIP1
Gene Name
MAPK regulated corepressor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093767.2 Intron NP_001087236.1
NM_001288798.1 Intron NP_001275727.1
NM_001288799.1 Intron NP_001275728.1
NM_207368.4 Intron NP_997251.2
XM_011523571.2 Intron XP_011521873.1
XM_011523572.2 Intron XP_011521874.1

View Full Product Details