Product Details

SNP ID
rs150321809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41501347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTCGTGGTAACAGAGGTGCTA[C/T]GGGGGCTGACGCTTCCTGGGAAACA
Phenotype
MIM: 148065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT13 PubMed Links

Gene Details

Gene
KRT13
Gene Name
keratin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002274.3 1323 Silent Mutation CCA,CCG P420P NP_002265.2
NM_153490.2 1323 Missense Mutation CAT,CGT H429R NP_705694.2

View Full Product Details