Product Details

SNP ID
rs150564817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81888531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCGGAAGTCACCTCTGTGCAGG[C/T]CTCCGCTGTGCGTCAATCTGTGACG
Phenotype
MIM: 604171 MIM: 614534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005782.3 597 Silent Mutation AGA,AGG R197R NP_005773.3
Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

View Full Product Details