Product Details

SNP ID
rs150627975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16441403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGATGTCAGTCCCAAGGACCA[C/G]TAGCTGCCATCAGTTTGGATTCTGA
Phenotype
MIM: 606676
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC75A PubMed Links

Gene Details

Gene
LRRC75A
Gene Name
leucine rich repeat containing 75A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113567.2 Intron NP_001107039.1
NM_207387.3 Intron NP_997270.2
XM_011523845.2 Intron XP_011522147.1
XM_017024619.1 Intron XP_016880108.1
XM_017024620.1 Intron XP_016880109.1
Gene
LRRC75A-AS1
Gene Name
LRRC75A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SNORD49A
Gene Name
small nucleolar RNA, C/D box 49A
There are no transcripts associated with this gene.

Gene
SNORD49B
Gene Name
small nucleolar RNA, C/D box 49B
There are no transcripts associated with this gene.

Gene
SNORD65
Gene Name
small nucleolar RNA, C/D box 65
There are no transcripts associated with this gene.

Gene
TRPV2
Gene Name
transient receptor potential cation channel subfamily V member 2
There are no transcripts associated with this gene.

View Full Product Details