Product Details

SNP ID
rs150774233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81665464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTTGGGAGCCCACCTCTACGT[C/G]GTCTGTCCCGAATTTTCTGCGCCCA
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC137 PubMed Links

Gene Details

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
There are no transcripts associated with this gene.

Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039842.2 241 Missense Mutation CAC,GAC H61D NP_001034931.1
NM_001304994.1 241 Missense Mutation CAC,GAC H51D NP_001291923.1
NM_001304995.1 241 UTR 3 NP_001291924.1
NM_001304999.1 241 UTR 3 NP_001291928.1
Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details