Product Details

SNP ID
rs150832197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29933289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTCTGGGCAAATGTGCTCTTTA[C/T]GATATTGGCTTTCAGGCTTTGTCTG
Phenotype
MIM: 606779
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCAB5 PubMed Links

Gene Details

Gene
EFCAB5
Gene Name
EF-hand calcium binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145053.1 Intron NP_001138525.1
NM_198529.3 Intron NP_940931.2
XM_011524759.1 Intron XP_011523061.1
XM_011524763.2 Intron XP_011523065.1
XM_011524765.1 Intron XP_011523067.1
XM_011524766.2 Intron XP_011523068.1
XM_011524768.2 Intron XP_011523070.1
XM_017024591.1 Intron XP_016880080.1
XM_017024592.1 Intron XP_016880081.1
Gene
SSH2
Gene Name
slingshot protein phosphatase 2
There are no transcripts associated with this gene.

View Full Product Details